missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Aluminum Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

$426.60 - $1070.00

Chemical Identifiers

CAS 13473-90-0
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RV010332100
View Documents
Ricca Chemical Company
RV010332100N
100 mL
Each for $426.60
Only null left
 
RV010332250
View Documents
Ricca Chemical Company
RV010332250N
250 mL
Each for $751.53
Only null left
 
RV010332500
View Documents
Ricca Chemical Company
RV010332500N
500 mL
Each for $1,070.00
Only null left
 
Description

Description

  • 1000 ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Chemical Identifiers

13473-90-0

Specifications

Natural LDPE Bottle
94.68%,1.96%,1.36%
1
Approximately 0°C
100 mL
13473-90-0 , 7697-37-2 , 7732-18-5
98.54%,2.04%,1.42%
Liquid
Standard
High Purity
Videos
SDS
Documents

Documents