missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Aluminum (Al) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

$246.03 - $433.43

Chemical Identifiers

CAS 13473-90-0
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RV010064100
View Documents
Ricca Chemical Company
RV010064100N
100 mL
Each for $246.03
Only null left
 
RV010064500
View Documents
Ricca Chemical Company
RV010064500N
500 mL
Each for $433.43
Only null left
 
Description

Description

  • 1000 ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Chemical Identifiers

13473-90-0

Specifications

Natural LDPE Bottle
94.68%,1.96%,1.36%
1.01
Approximately 0°C
High Purity
13473-90-0 , 7697-37-2 , 7732-18-5
98.54%,2.04%,1.42%
Liquid
100 mL
Videos
SDS
Documents

Documents