missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Aluminum (Al) Standard for AAS 1000 ppm in 2% HCl, Ricca Chemical

Catalog No. RV010002100
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010002100 100 mL
RV010002500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010002100 Supplier Ricca Chemical Company Supplier No. RV010002100N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Chemical Identifiers

CAS 7784-27-2 , 7647-01-0 , 7446-70-0 , 7732-18-5
Molecular Formula AlH18N3O18
Molecular Weight (g/mol) 375.129
InChI Key SWCIQHXIXUMHKA-UHFFFAOYSA-N
PubChem CID 24567
IUPAC Name aluminum;trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]

Specifications

Packaging Natural LDPE Bottle
CAS Min % 0.955598, 0.01%, 0.004802
CAS Max % 99.4602, 0.0204, 0.004998
Specific Gravity 1.013
Physical Form Liquid
Melting Point Approximately 0°C
Type Standard
Industry Type Atomic Absorption
Quantity 100 mL
Grade High Purity
Chemical Name or Material Aluminum (Al) Standard for AAS
Show More Show Less