missing translation for 'onlineSavingsMsg'
Learn More

Veratrole, 99+%, Thermo Scientific™

Catalog No. AC161111000
Change view
Click to view available options
Quantity:
100g
500g
1kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161111000 100g Glass bottle
AC161115000 500g Glass bottle
AC161110010 1kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC161111000 Supplier Thermo Scientific Chemicals Supplier No. 161111000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 91-16-7
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00008357
InChI Key ABDKAPXRBAPSQN-UHFFFAOYSA-N
Synonym veratrole, veratrol, pyrocatechol dimethyl ether, o-dimethoxybenzene, catechol dimethyl ether, benzene, 1,2-dimethoxy, 2-methoxyanisole, o,o-dimethyl catechol, 2-dimethoxybenzol, benzene, o-dimethoxy
PubChem CID 7043
ChEBI CHEBI:59114
IUPAC Name 1,2-dimethoxybenzene
SMILES COC1=CC=CC=C1OC

Specifications

Density 1.0800g/mL
Boiling Point 206°C to 207°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5330 to 1.535
Linear Formula C6H4(OCH3)2
Quantity 100g
Beilstein 06, 771
Specific Gravity 1.08
Merck Index 15, 10150
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,methanol,acetone and ether
Formula Weight 138.17
Percent Purity 99+%
Chemical Name or Material Veratrole, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 202-045-3
RTECSNumber CZ6475000
TSCA TSCA

RUO – Research Use Only