Learn More
MSA™ Ventilation Smoke Tube Kits
Description
- Allows for controlled generation of visible smoke
- Determines velocity and flow patterns of heating, ventilating and air-conditioning systems
- Available with glass or plastic tubes with break-off tips
Use these kits to determine the velocity of slow-moving air currents and establish their direction and flow patterns. MSA™ Ventilation Smoke Tube Kits are ideal for use in commercial buildings and industrial processing plants as well as in shafts, mines and tunnels.
- Allows for controlled generation of visible smoke
- Determines velocity and flow patterns of heating, ventilating and air-conditioning systems
- Available with glass or plastic tubes with break-off tips
Specifications
Specifications
| Description | Ventilation Smoke Kit, including aspirator bulb, two rubber plugs, and six smoke-producing tubes contained in plastic carrying case |
| Type | Smoke Tube Kit |
| Material | Glass |
| Quantity | 1 Kit |
| Tip Type | Break-off |