Learn More
New England Biolab Ltd Vent (exo-) DNA Polymerase 200 units
Numéro de catalogue. 50811677
Encompass
Vent (exo-) DNA Polymerase has been genetically engineered to eliminate the 3 5 proofreading exonuclease activity associated with Vent DNA Polymerase. This is the preferred form for high-temperature dideoxy sequencing reactions and for high yield primer extension reactions. The fidelity of polymerization by this form is reduced to a level about 2-fold higher than that of Taq DNA Polymerase.
Numéro de catalogue. 50811677 Fournisseur New England Biolab Ltd Code fournisseur M0257S
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Vent (exo-) DNA Polymerase has been genetically engineered to eliminate the 3 5 proofreading exonuclease activity associated with Vent DNA Polymerase. This is the preferred form for high-temperature dideoxy sequencing reactions and for high yield primer extension reactions. The fidelity of polymerization by this form is reduced to a level about 2-fold higher than that of Taq DNA Polymerase.