missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Venlafaxine hydrochloride

CAS: 99300-78-4 | C17H28ClNO2, C17H27NO2·HCl | 313.87 g/mol

$296.65 - $296.65

Chemical Identifiers

CAS 99300-78-4
Molecular Formula C17H28ClNO2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC465810050
View Documents
Thermo Scientific Chemicals
465810050
5 g
Each for $296.65
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

99300-78-4
C17H28ClNO2

Specifications

99300-78-4
Solubility aq. soln.: slightly soluble.
C17H28ClNO2
QYRYFNHXARDNFZ-UHFFFAOYNA-N
hydrogen 1-[2-(dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol chloride
Venlafaxine hydrochloride
5 g
217.0°C
313.86
[H+].[Cl-].COC1=CC=C(C=C1)C(CN(C)C)C1(O)CCCCC1
313.87
Videos
Safety and Handling

Safety and Handling

GHS H Statement Causes serious eye irritation.

GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

Wear eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only