missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Venlafaxine hydrochloride

Catalog No. AC465810010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465810010 1 g
AC465810050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465810010 Supplier Thermo Scientific Chemicals Supplier No. 465810010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 99300-78-4 | C17H28ClNO2, C17H27NO2·HCl | 313.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99300-78-4
Molecular Formula C17H28ClNO2

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: slightly soluble.
Melting Point 217.0°C
Formula Weight 313.86
InChI Key QYRYFNHXARDNFZ-UHFFFAOYNA-N
SMILES [H+].[Cl-].COC1=CC=C(C=C1)C(CN(C)C)C1(O)CCCCC1
IUPAC Name hydrogen 1-[2-(dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol chloride
Molecular Weight (g/mol) 313.87
Chemical Name or Material Venlafaxine hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes serious eye irritation.

Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

Wear eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only