missing translation for 'onlineSavingsMsg'
Learn More

Veliparib, Thermo Scientific™

Catalog No. AC469261000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469261000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC469261000 Supplier Thermo Scientific Chemicals Supplier No. 469261000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 912444-00-9
Molecular Formula C13 H16 N4O

Specifications

Quantity 100 mg
Formula Weight 244.30
Physical Form Powder
Color White to Yellow
Purity ≥98.5%
Chemical Name or Material Veliparib
Hazard Category Exclamation mark
Recommended Storage Normal conditions