Learn More
Vector Laboratories VECTASTAIN ABC-AmP Reagent PK-4007
Numéro de catalogue. 502045694
Encompass
VECTASTAIN ABC-AmP Reagent (Standard, Western Blot Detection), 1 Kit, is an amplified ABC alkaline phosphatase reagent for the detection of mouse or rabbit primary antibodies on nitrocellulose or PVDF membranes. The signal can be visualized using either a chemiluminescent/chemifluorescent or a chromogenic substrate. Where a DuoLuX Chemiluminescent and Fluorescent Substrate (SK-6605-100) is used, VECTASTAIN ABC-AmP produces a very high and sustained light emission signal with low background and permanent fluorescence, allowing for the detection of as little as 1 pg of target protein. VECTASTAIN ABC-AmP Reagent is sufficient to develop approximately 20 x 100 cm2 blots.
Numéro de catalogue. 502045694 Fournisseur Vector Laboratories Code fournisseur PK4007
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
VECTASTAIN ABC-AmP Reagent (Standard, Western Blot Detection), 1 Kit, is an amplified ABC alkaline phosphatase reagent for the detection of mouse or rabbit primary antibodies on nitrocellulose or PVDF membranes. The signal can be visualized using either a chemiluminescent/chemifluorescent or a chromogenic substrate. Where a DuoLuX Chemiluminescent and Fluorescent Substrate (SK-6605-100) is used, VECTASTAIN ABC-AmP produces a very high and sustained light emission signal with low background and permanent fluorescence, allowing for the detection of as little as 1 pg of target protein. VECTASTAIN ABC-AmP Reagent is sufficient to develop approximately 20 x 100 cm2 blots.