VECTASTAIN Elite ABC Kit (Universal),1 kit,3 ml normal serum,2 ml of concentrated universal biotinylated secondary antibody (for mouse and rabbit primary antibodies),and 2 ml each of concentrated A and B reagents
Numéro de catalogue.
NC9365554
Fournisseur
Vector LaboratoriesCode fournisseur
PK6200
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
VECTASTAIN Elite ABC Kit (Universal),1 kit,3 ml normal serum,2 ml of concentrated universal biotinylated secondary antibody (for mouse and rabbit primary antibodies),and 2 ml each of concentrated A and B reagents