Learn More
Molecular Toxicology Inc 50X VBE Salts - Sterile, 140 mL/bottle, Store at Room Temperature
Numéro de catalogue. 50948746
Encompass
Vogel-Bonner medium E (VBE) and Vogel""s salts are used in the manufacture of Minimal Glucose Agar for use in the Ames assay. MOLTOX offers a wide array of solutions and buffers for a variety of laboratory research needs. All reagents are accompanied by a GLP-compliant Formulation and Quality Control statement.
Numéro de catalogue. 50948746 Fournisseur Molecular Toxicology Inc Code fournisseur 26541
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Vogel-Bonner medium E (VBE) and Vogel""s salts are used in the manufacture of Minimal Glucose Agar for use in the Ames assay. MOLTOX offers a wide array of solutions and buffers for a variety of laboratory research needs. All reagents are accompanied by a GLP-compliant Formulation and Quality Control statement.