Learn More
Biomatik Corporation Recombinant Vascular Endothelial Growth Factor B (VEGFB), 100UG
Numéro de catalogue. 501505775
Encompass
Recombinant Rat Vascular Endothelial Growth Factor B (VEGFB) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Vascular Endothelial Growth Factor B (VEGFB). Target Synonyms: VEGF-B; VEGFL; VRF; VEGF-Related Factor. Accession Number: O35485. Gene ID: 89811. Expression Region: Ala21~Ser137. Tag Info: N-terminal His Tag. Theoretical MW: 17kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Numéro de catalogue. 501505775 Fournisseur Biomatik Corporation Code fournisseur RPU44324
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant Rat Vascular Endothelial Growth Factor B (VEGFB) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Vascular Endothelial Growth Factor B (VEGFB). Target Synonyms: VEGF-B; VEGFL; VRF; VEGF-Related Factor. Accession Number: O35485. Gene ID: 89811. Expression Region: Ala21~Ser137. Tag Info: N-terminal His Tag. Theoretical MW: 17kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.