Learn More
Cole-Parmer™ VapLock™ Solvent Waste Kits for Bottles and Carboys
Description
Various cap sizes available to fit your solvent container
Warranty
None
Specifications
Specifications
| For Use With (Equipment) | Solvent-Compatible Containers |
| Material | Polypropylene |
| Size | GL38 |
| Thread Size | 1 x 1/2 in. NPT(F), 3 x 1/4 in. NPT(F), 6 x 1/4 in.-28 UN(F)(F) |
| For Use With (Application) | Solvent Safety |
| Product Type | Solvent Waste Kit |