missing translation for 'onlineSavingsMsg'
Learn More

Vanillyl Alcohol 98.0+%, TCI America™

Catalog No. V001825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V001825G 25 g
V0018500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V001825G Supplier TCI America Supplier No. V001825G

Chemical Identifiers

CAS 498-00-0
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00004659
InChI Key ZENOXNGFMSCLLL-UHFFFAOYSA-N
Synonym vanillyl alcohol, 4-hydroxy-3-methoxybenzyl alcohol, 4-hydroxymethyl-2-methoxyphenol, vanillin alcohol, vanillic alcohol, 4-hydroxy-3-methoxybenzenemethanol, benzenemethanol, 4-hydroxy-3-methoxy, vanillol, 4-hydroxy-3-methoxybenzylalcohol, 4-hydroxy-3-methoxyphenylmethanol
PubChem CID 62348
ChEBI CHEBI:18353
IUPAC Name 4-(hydroxymethyl)-2-methoxyphenol
SMILES COC1=CC(CO)=CC=C1O

Specifications

Melting Point 114°C
Color White-Yellow
Quantity 25 g
Formula Weight 154.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Vanillyl Alcohol