missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanadium(V) oxytriethoxide, 95%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
Spécifications
| Nom ou substance chimique | Vanadium(V) oxytriethoxide |
| Densité | 1.1390g/mL |
| Point d’ébullition | 125.0°C to 128.0°C |
| Point d’éclair | 71°C |
| Spectre infrarouge | Authentic |
| Fourchette de pourcentages d’essai | 95% |
| Emballage | Glass bottle |
| Indice de réfraction | 1.5080 to 1.5120 |
| Formule linéaire | (CH3CH2O)3V=O |
| Quantité | 5mL |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. If eye irritation persists: Get medical advice/attention. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER |
| EINECSNumber | 216-870-1 |
RUO – Research Use Only