missing translation for 'onlineSavingsMsg'
Learn More

Spectrum Chemical Vanadium Pentoxide, Powder, 99.5%, Spectrum™ Chemical

Catalog No. 18614644
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614644 25 g Amber Glass Bottle
18614639 125 g Amber Glass Bottle
18614641 500 g Amber Glass Bottle
18614643 2.5 kg Amber Glass Bottle
18614640 12 kg Poly Pail
18614642 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18614644 Supplier Spectrum Chemical Supplier No. V101025GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • Vanadium Pentoxide, Powder, Reagent is an inorganic compound that is a brown to yellow solid that is both an amphoteric oxide and an oxidizing agent.
  • It is used as the main component in the production of ferrovanadium, a key component in the production of carbon steel used for infrastructure, and an important one in the production of sulfuric acid.
  • Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 1314-62-1

Specifications

Boiling Point 1750°C
Melting Point 690°C
Color Yellow-orange to Yellowish-red
CAS Max % 1
Physical Form Solid
Quantity 25 g
Assay Percent Range 99.5%
Molecular Formula V2O5
Linear Formula V2O5
UN Number UN2862
Synonym Vanadic Anhydride
Solubility Information Soluble in Acetone, Partially Soluble in Cold Water, Soluble in Acid
IUPAC Name Divanadium Pentaoxide, Divanadium Pentoxide, Pentaoxodivanadium, Vanadic Anhydride, Vanadic Acid Anhydride, Vanadium (V) Oxide, Vanadium Pentaoxide
Molecular Weight (g/mol) 181.9
Formula Weight 181.90 g/mol
Percent Purity 99.5%
Grade Reagent
Manufacturing Origin USA
Odor Odorless
Packaging Amber Glass Bottle
Appearance Powder
Specific Gravity 3.357
Chemical Name or Material Vanadium Pentoxide
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.