missing translation for 'onlineSavingsMsg'
Learn More

Vanadium ICP Standard, 1000 ppm V in 3% HNO3, Ricca Chemical

Catalog No. PV1KN500
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PV1KN500 500 mL Natural Poly Bottle
PV1KN100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PV1KN500 Supplier Ricca Chemical Company Supplier No. PV1KN500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1314-62-1 , 7697-37-2 , 7732-18-5
Molecular Formula O5V2
Molecular Weight (g/mol) 181.88
MDL Number MFCD00011457
InChI Key GNTDGMZSJNCJKK-UHFFFAOYSA-N
PubChem CID 14814
ChEBI CHEBI:30045
IUPAC Name [(dioxovanadio)oxy]dioxovanadium
SMILES O=[V](=O)O[V](=O)=O

Specifications

Packaging Natural Poly Bottle
CAS Min % 92.0, 5.87, 0.16
CAS Max % 96.0, 6.11, 0.17
Physical Form Liquid
Color Yellow
pH < 2
Melting Point Approximately 0°C
Type Standard
Quantity 500 mL
Concentration or Composition (by Analyte or Components) 1mL = 1mg V (1, 000ppm V)V2O5 in 3% HNO3
Chemical Name or Material Vanadium ICP Standard
Show More Show Less
ShelfLife 18 months