missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanadium AA Standard, 1000 ppm V in 8% HCl, Ricca Chemical
$116.13 - $116.13
Chemical Identifiers
| CAS | 1314-62-1 |
|---|---|
| Molecular Formula | O5V2 |
| Molecular Weight (g/mol) | 181.88 |
| MDL Number | MFCD00011457 |
| InChI Key | GNTDGMZSJNCJKK-UHFFFAOYSA-N |
| PubChem CID | 14814 |
| ChEBI | CHEBI:30045 |
| IUPAC Name | [(dioxovanadio)oxy]dioxovanadium |
| SMILES | O=[V](=O)O[V](=O)=O |
Chemical Identifiers
| 1314-62-1 | |
| 181.88 | |
| GNTDGMZSJNCJKK-UHFFFAOYSA-N | |
| CHEBI:30045 | |
| O=[V](=O)O[V](=O)=O |
| O5V2 | |
| MFCD00011457 | |
| 14814 | |
| [(dioxovanadio)oxy]dioxovanadium |
Specifications
| Natural LDPE Bottle | |
| 82.0,15.54,0.15 | |
| Odorless | |
| MFCD00011457 | |
| Approximately 0°C | |
| GNTDGMZSJNCJKK-UHFFFAOYSA-N | |
| [(dioxovanadio)oxy]dioxovanadium | |
| 100 mL | |
| CHEBI:30045 | |
| Vanadium AA Standard |
Safety and Handling
ShelfLife : 24 months