missing translation for 'onlineSavingsMsg'
Learn More

Valeric Acid 98.0+%, TCI America™

Catalog No. V0003500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0003500ML 500 mL
V000325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V0003500ML Supplier TCI America Supplier No. V0003500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-52-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00004413
InChI Key NQPDZGIKBAWPEJ-UHFFFAOYSA-N
Synonym valeric acid, n-valeric acid, n-pentanoic acid, valerianic acid, 1-butanecarboxylic acid, propylacetic acid, butanecarboxylic acid, pentoic acid, kyselina valerova, valeric acid, n
PubChem CID 7991
ChEBI CHEBI:17418
IUPAC Name pentanoic acid
SMILES CCCCC(=O)O

Specifications

Melting Point -18°C
Color Yellow
Boiling Point 186°C
Quantity 500 mL
UN Number 3265
Formula Weight 102.13
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Valeric Acid
EINECSNumber (2)-0604
RTECSNumber YV6100000
TSCA Yes