missing translation for 'onlineSavingsMsg'
Learn More

Valeric acid hydrazide, 95%

Catalog No. AAL0155514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0155514 25 g
AAL0155506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0155514 Supplier Thermo Scientific Chemicals Supplier No. L0155514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 38291-82-6 | C5H12N2O | 116.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38291-82-6
Molecular Formula C5H12N2O
Molecular Weight (g/mol) 116.164
MDL Number MFCD00025137
InChI Key PJBQYCIDGYKEMN-UHFFFAOYSA-N
Synonym valeric acid hydrazide, pentanohydrazide, valerohydrazide, pentanoic acid, hydrazide, pentanoic acid hydrazide, valerylhydrazine, pentanoic acid,hydrazide, acmc-1afhr
PubChem CID 93202
IUPAC Name pentanehydrazide
SMILES CCCCC(=O)NN

Specifications

Melting Point 55°C to 64°C
Quantity 25 g
Beilstein 1744747
Formula Weight 116.16
Percent Purity 95%
Chemical Name or Material Valeric acid hydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 253-864-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only