Learn More
Valco™ Precision Sampling Syringe, Series A-2, Sold by MilliporeSigma™ Supelco™
An economically priced general purpose syringe, it is leak-tight to 250 psi for both gas and liquids
Supplier:  Merck Emd Millipore SGMA22272
 View more versions of this product
Catalog No. 111010732
Only null left
Description
Description
  • Push-button valve allows instantaneous injection
  • Great for small-volume, volatile samples
Specifications
Chemical Identifiers
Stainless Steel (Tip)
2 in.
2
Valco
Removable
Series A-2
Precision Sampling Syringe
1 Ea.
Bevel
Specifications
Stainless Steel (Tip)
0.012 in.
2 in.
250 psi
1 Ea.
Valco
Removable
Series A-2
0.029 in.
Precision Sampling Syringe
2
500 μL
Bevel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Valco is a registered trademark of Valco Instruments Co. Inc.