missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valco™ Precision Sampling Syringe, Series A-2, Sold by MilliporeSigma™ Supelco™
An economically priced general purpose syringe, it is leak-tight to 250 psi for both gas and liquids
Supplier: Merck Emd Millipore 22271
Description
- Push-button valve allows instantaneous injection
- Great for small-volume, volatile samples
Chemical Identifiers
| Stainless Steel (Tip) | |
| 2 in. | |
| 2 | |
| Valco | |
| Removable |
| Series A-2 | |
| Precision Sampling Syringe | |
| 1 Ea. | |
| Bevel |
Specifications
| Stainless Steel (Tip) | |
| 0.005 in. | |
| 2 in. | |
| 2 | |
| 250 μL | |
| Bevel |
| Series A-2 | |
| 0.028 in. | |
| Precision Sampling Syringe | |
| 1 Ea. | |
| Valco | |
| Removable |
Valco is a registered trademark of Valco Instruments Co. Inc.