missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Vacuum Seal

Numéro de catalogue. p-7222745
Change view
Click to view available options
Includes:
FFKM Sealing Ring
FKM Sealing Ring
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Includes
02321080 FKM Sealing Ring
02321108 FFKM Sealing Ring
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 02321080 Fournisseur Heidolph™ Code fournisseur 036210032
Il en reste null

The highly durable and long lasting PTFE vacuum seals with an FKM or an FFKM sealing ring to achieve maximum tightness and save on long-term spare parts costs.

  • PTFE spare seal
  • Choose between an FKM or an FFKM sealing ring
  • Materials offer very good chemical resistance and the high quality sealing properties
  • Long running time without abrasion
  • Highest density
  • FDA compliant

Spécifications

Color Natural
Material PTFE
Type Vacuum Seal
For Use With (Equipment) Evaporators
Includes FKM Sealing Ring