Learn More
BINDER™ Vacuum Module with Vacuum Pump VP 2
Vacuum module with vacuum pump VP 2 - 120 V / 50-60 Hz option model, Pump chamber for installation of vacuum pumps, with switchable power socket.
Supplier: BINDER™ 80122619
Description
- Temperature range:
- +9°C above ambient temperature to +110 °C
- Controller with digital display of pressure and temperature
- Program-controlled drying monitoring with automatic ventilation at end of process
- Internal data logger, measured values can be read out in open format via USB
- 1 aluminum expansion rack, can be custom-positioned
- Inert gas connection
- Universal access port DN 16 - for the introduction of external measuring devices
- Shatterproof, spring-mounted safety glass panel
- Large viewing window with option of interior lighting
Identifiants chimiques
| 50/60 Hz | |
| 581 x 743 x 942 mm | |
| 110°C | |
| 120 V |
Spécifications
| 3 to 5.7 | |
| 581 x 743 x 942 mm | |
| BINDER units VD023UL-120V |
| 50/60 Hz | |
| 110°C | |
| 120 V |