Learn More
Synthware™ Vacuum Manifold with PTFE Stopcocks, Left/Right Side Connection
GREENER_CHOICE
Catalog No. 31501783
Change view
Click to view available options
Port Size (Metric):
2 mm Stopcock Bore
4 mm Stopcock Bore
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Port Size (Metric)
31501783 4 mm Stopcock Bore
31501782 2 mm Stopcock Bore
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31501783 Supplier Synthware™ Supplier No. M490064
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
PTFE stopcocks eliminate the need for grease.
Main tube is 22 mm O.D. with serrated hose connection (10 mm O.D.) and delivery ports have 10 mm serrated hose connections. Distance between stopcocks is 75 mm. -Hose connection : Both Sides
Specifications
Connection Type Left and Right Hose Connection
Type Vacuum Manifold
For Use With (Application) Organic Synthesis
Includes PTFE Stopcocks
Material Borosilicate Glass
No. of Ports 6
Port Size (Metric) 4 mm Stopcock Bore