Learn More
Chemglass Inc Vacuum/Inert Manifold, 4 Ports, 500mm Overall Width
Numéro de catalogue. CG443602
Encompass
Vacuum/Inert Manifold, 4 Ports, 500mm Overall Width. General purpose double bank vacuum manifold with 4 ports for use with vacuum and inert gas. Both tubes are 1/2 OD and have the upper left and lower right banks open. Ports are 0-4mm Chem-Vac™ Chem-Cap valves and are spaced 100mm center to center with 9mm OD tubulation.
Numéro de catalogue. CG443602 Fournisseur Chemglass Inc Code fournisseur CG443602
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Vacuum/Inert Manifold, 4 Ports, 500mm Overall Width. General purpose double bank vacuum manifold with 4 ports for use with vacuum and inert gas. Both tubes are 1/2 OD and have the upper left and lower right banks open. Ports are 0-4mm Chem-Vac™ Chem-Cap valves and are spaced 100mm center to center with 9mm OD tubulation.