Learn More
Thermo Scientific™ Vacuum Hydrolysis Tube
Catalog No. TS29571
Change view
Click to view available options
Capacity (Metric):
6 mL
Catalog No. Capacity (Metric)
TS29571 6 mL
1 options
Catalog No. TS29571 Supplier Thermo Scientific™ Supplier No. TS29571
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Perform derivatization and other small-scale reactions reliably and with ease.
Ideal applications
  • General sample incubation and evaporation in a variety of tube and vial sizes
  • Silylation, alkylation, and acylation derivatization reactions for GC sample preparation
  • Protein hydrolysis and vacuum hydrolysis reactions for amino acid analysis by HPLC
Additional features
  • LED display: match digital display to in-block thermometer to calibrate temperature set-point
  • Modular design: switch aluminum blocks and vials; attach compatible evaporator manifold
  • Four models: single-block and triple-block sizes with either heat-only or heat-and-stir capability
  • In-block temperature control option by Remote Temperature Probe, an optional accessory to allow temperature regulation from block wells or actual sample vials
Complete Reacti-Therm Sample Derivatization System
The derivatization system can combine heating, stirring and evaporation in a versatile, modular system that is easily configured to your application.
  • Reacti-Therm Heating/Stirring Module
  • Reacti-Vap Evaporator Module
  • Reacti-Blocks, Reacti-Vial Magnetic Stirrers, and Reacti-Vial Small Reaction Vials
Specifications
Capacity (Metric) 6 mL
Quantity 1