Learn More
Greiner Bio-One™ VACUETTE™ SAFELINK holder
Description
- Sterile, single-use tube holder with male luer lock.
- Easy to use: Suitable for all standardized female luer lock connections.
- Safe sampling with secure, threaded luer lock connection.
- Suitable for pressures up to 185 mmHg.
- Not made with natural rubber latex.
- Makes SAFELINK a practical accessory for blood collection.
Spécifications
Spécifications
| Matériau | Polypropylene |
| Autoclavable | Not Autoclavable |
| Couleur | Transparent |
| Type de produit | Blood Transfer Unit |
| Jetable | Single-use |
| Sans latex | Yes |
| Stérilité | Sterile |