Learn More
Greiner Bio-One VACUETTE™ K3EDTA Blood Collection Tubes
VACUETTE™ EDTA Blood Collection tubes are used for determinations in EDTA whole blood for haematology. Available with K2EDTA, K2EDTA Separator and K3EDTA. VACUETTE™ K3EDTA tubes contain tri-potassium K3EDTA which blocks the coagulation cascade.
$25.24 - $38.55
Specifications
| Product Type | Blood Collection Tube |
|---|---|
| Additive | K3 EDTA |
| Material | PET |
| Sterility | Sterile |
| For Use With (Equipment) | VACUETTE™ Blood Collection Needles |
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Color | Diameter (Metric) Outer | Closure Type | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Color | Diameter (Metric) Outer | Closure Type | Price | Quantity & Availability | ||
|
22040035
|
Greiner Bio-One
454217 |
3 mL | Lavender with Black Ring | 13 mm | Safety Pull Cap |
Pack of 50 Each for $25.24
Case of 24 Pack for $605.76
|
|
||
|
22040162
|
Greiner Bio-One
GBO456003 |
6 mL | Pink with Black Ring | 13 x 100 mm | Safety Pull Cap |
Pack of 50 Each for $28.38
Case of 24 Pack for $681.12
|
|
||
|
22040096
|
Greiner Bio-One
456067 |
6 mL | Pink with Black Ring | 13 mm | Safety Pull Cap |
Pack of 50 Each for $34.17
Case of 24 Pack for $820.08
|
|
||
|
22040101
|
Greiner Bio-One
454222 |
2 mL | Lavender with White Ring | 13 mm | Safety Pull Cap |
Pack of 50 Each for $25.38
Case of 24 Pack for $609.12
|
|
||
|
22040037
|
Greiner Bio-One
455036 |
9 mL | Lavender with Black Ring | 16 mm | Safety Twist Cap |
Pack of 50 Each for $38.55
Case of 24 Pack for $925.20
|
|
||
|
22040129
|
Greiner Bio-One
456038 |
6 mL | Lavender with Black Ring | 13 mm | Safety Pull Cap |
Pack of 50 Each for $27.03
Case of 24 Pack for $648.72
|
|
||
|
22040036
|
Greiner Bio-One
454021 |
4 mL | Lavender with Black Ring | 13 mm | Safety Pull Cap |
Pack of 50 Each for $32.76
Case of 24 Pack for $786.24
|
|
||
Description
VACUETTE™ EDTA Blood Collection tubes are used for determinations in EDTA whole blood for haematology. Available with K2EDTA, K2EDTA Separator and K3EDTA. VACUETTE™ K3EDTA tubes contain tri-potassium K3EDTA which blocks the coagulation cascade.
- Tubes are supplied in packs of 50 pieces
- For venous blood collection
- Made out of virtually unbreakable PET plastic
- Improved safety and hygiene during specimen collection procedures with the advantage of being as clear as glass
- Primarily in hematology
- Inner wall is coated with spray-dried K2EDTA which acts as an anticoagulant, binding calcium ions and interrupting the clotting cascade
- K2EDTA Blood Collection Tubes with Gel Separator and K3EDTA Blood Collection Tubes are available
- Pink caps for immunohematology are also available
- CE marked
Specifications
| Blood Collection Tube | |
| PET | |
| VACUETTE™ Blood Collection Needles |
| K3 EDTA | |
| Sterile | |
| Blood Collection |