Learn More
Greiner Bio-One VACUETTE™ K3EDTA Blood Collection Tubes
VACUETTE™ EDTA Blood Collection tubes are used for determinations in EDTA whole blood for haematology. Available with K2EDTA, K2EDTA Separator and K3EDTA. VACUETTE™ K3EDTA tubes contain tri-potassium K3EDTA which blocks the coagulation cascade.
Supplier:  Greiner Bio-One 456067
 View more versions of this product
Catalog No. 22040096
Only null left
Description
Description
VACUETTE™ EDTA Blood Collection tubes are used for determinations in EDTA whole blood for haematology. Available with K2EDTA, K2EDTA Separator and K3EDTA. VACUETTE™ K3EDTA tubes contain tri-potassium K3EDTA which blocks the coagulation cascade.
  • Tubes are supplied in packs of 50 pieces
  • For venous blood collection
  • Made out of virtually unbreakable PET plastic
  • Improved safety and hygiene during specimen collection procedures with the advantage of being as clear as glass
  • Primarily in hematology
  • Inner wall is coated with spray-dried K2EDTA which acts as an anticoagulant, binding calcium ions and interrupting the clotting cascade
  • K2EDTA Blood Collection Tubes with Gel Separator and K3EDTA Blood Collection Tubes are available
  • Pink caps for immunohematology are also available
  • CE marked
Specifications
Chemical Identifiers
Blood Collection Tube
PET
VACUETTE™ Blood Collection Needles
K3 EDTA
Sterile
Blood Collection
Specifications
Blood Collection Tube
PET
Safety Pull Cap
VACUETTE™ Blood Collection Needles
100 mm
Blood Collection
K3 EDTA
Sterile
6 mL
Pink with Black Ring
13 x 100 mm
13 mm
Product Suggestions
Product Suggestions