Learn More
Foxx Life Sciences VactrapXL™ Vacuum Trap System (Vacuum Flask) for Aspiration and Vacuum Protection, Shatterproof 4+2 Liter Aspirator Bottle Assembly with Secondary Container, Autoclavable Polypropylene (PP), 4L Bottle with 2L Overflow Bottle and Inline Vent Filter
GREENER_CHOICE
Numéro de catalogue. 501720651
Encompass
Foxx Life Sciences VactrapXL™ is the world's most advanced vacuum trap system. Ideal for protection of vacuum lines and scientists alike, VactrapXL safely contains waste from supernatural removal, chemical separation, and cell or tissue culture media aspiration. Best of all, VactrapXL™ comes ready to use; simply attach to your vacuum and begin your work. The full VactrapXL™ system includes two Foxx Heavy Duty Vacuum Bottles (autoclavable PP or chemically-resistant HDPE), each with a patented VersaCap® closure for tangle-free opening, no need to disconnect tubing, ever. A 0.2m PTFE inline vent filter retains any aerosolized bacteria, and the entire assembly is nested neatly in a secondary container with a small footprint and incredible stability. A pinch clamp allows for modulation of vacuum. Other Vactrap options offer one bottle, or setups without secondary containment. Say goodbye to glass. Say hello to Vactrap.
Numéro de catalogue. 501720651 Fournisseur Foxx Life Sciences Code fournisseur 3054201FLS
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Foxx Life Sciences VactrapXL™ is the world's most advanced vacuum trap system. Ideal for protection of vacuum lines and scientists alike, VactrapXL safely contains waste from supernatural removal, chemical separation, and cell or tissue culture media aspiration. Best of all, VactrapXL™ comes ready to use; simply attach to your vacuum and begin your work. The full VactrapXL™ system includes two Foxx Heavy Duty Vacuum Bottles (autoclavable PP or chemically-resistant HDPE), each with a patented VersaCap® closure for tangle-free opening, no need to disconnect tubing, ever. A 0.2m PTFE inline vent filter retains any aerosolized bacteria, and the entire assembly is nested neatly in a secondary container with a small footprint and incredible stability. A pinch clamp allows for modulation of vacuum. Other Vactrap options offer one bottle, or setups without secondary containment. Say goodbye to glass. Say hello to Vactrap.