missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Utility Trays for Shaker Platforms
Description
Use these Thermo Scientific™ Utility trays for Shaker Platforms with 0.5 in. rubber mats.
Mix and match Thermo Scientific™ Solaris shaker Universal clamps to suit individual needs in the laboratory. Compatible with Solaris Open Air, Solaris Temperature Controlled Benchtop Shakers, MaxQ6000 Shakers.
Spécifications
Spécifications
| Product Type | Utility Tray |
| Dimensions (L x W) | 353 x 272 mm |
| Dimensions (LxW) Platform | 11 x 14 in. and 12 x 14 in. |
| Description | Utility Tray for Platforms |
| For Use With (Equipment) | Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers |