missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP17 Polyclonal Antibody

Description
USP17 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Deubiquitinating enzyme that removes conjugated ubiquitin from specific proteins to regulate different cellular processes that may include cell proliferation, progression through the cell cycle, apoptosis, cell migration, and the cellular response to viral infection. [UniProt].
Spécifications
Spécifications
| Antigen | USP17 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | USP17L24 |
| Gene Accession No. | Q0WX57 |
| Gene Alias | Deubiquitinating enzyme 17; RS447; Ubiquitin carboxyl-terminal hydrolase 17-like protein 24; ubiquitin specific peptidase 17-like family member 24; ubiquitin specific peptidase 17-like family member 9, pseudogene; Ubiquitin thioesterase 17; Ubiquitin-specific-processing protease 17; USP17; USP17A; USP17H; USP17I; USP17J; USP17K; USP17L; USP17L24; USP17L25; USP17L26; USP17L27; USP17L28; USP17L29; USP17L30; USP17L9P; USP17M |
| Gene Symbols | USP17L24 |
| Afficher plus de résultats |