Learn More
Uridine-5'-Diphosphoglucose Disodium Salt , MP Biomedicals™
Catalog No. MP210120880
Change view
Click to view available options
Quantity:
100 mg
Catalog No. Quantity
MP210120880 100 mg
1 options
Catalog No. MP210120880 Supplier MP Biomedicals Inc Supplier No. 0210120880
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Uridine-5'-diphosphoglucose Disodium Salt is a biosynthetic product produced by the reaction of UTP and glucose-1-phosphate catalyzed by uridyl transferase.
  • Endogenous nucleotide sugar involved in glycosyltransferase reactions in metabolism
  • Shown to bind the P2Y14 receptor, an atypical P2Y receptor involved in the activation of dendritic cells and glial cells
  • Binds to and activates GPR17, inducing oligodendrocyte differentiation at a maximal concentration of 100μM
Specifications
Quantity 100 mg