missing translation for 'onlineSavingsMsg'
Learn More

Urea, Shotted Pellets, Reagents

Numéro de catalogue. 01336899
Change view
Click to view available options
Quantity:
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
01336899 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 01336899 Fournisseur Labchem Inc Code fournisseur C23455002.5A5
Il en reste null

Urea, Shotted Pellets, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Identifiants chimiques

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.06
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
IUPAC Name urea
SMILES NC(N)=O

Spécifications

Boiling Range 135°C
Melting Point 133°C
Color White
Boiling Point 135°C
Melting Range 133°C
Packaging Poly Bottle
Quantity 2.5 kg
Specific Gravity 1.335
Grade Technical
Physical Form Solid
Chemical Name or Material Urea, Shotted Pellets
Afficher plus de résultats Afficher moins de résultats