missing translation for 'onlineSavingsMsg'
Learn More

Urea Peroxide, Spectrum™ Chemical

Catalog No. 18614631
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614631 500 g Amber Glass Bottle
18614628 100 g Amber Glass Bottle
18614629 2.5 kg Poly Pail
18614630 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18614631 Supplier Spectrum Chemical Mfg Cor Supplier No. UR110500GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 124-43-6

Specifications

CAS Max % 1
Melting Point 88° to 93°C
Packaging Amber Glass Bottle
Quantity 500 g
Grade Reagent

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.