missing translation for 'onlineSavingsMsg'
Learn More

Urea hydrogen peroxide adduct, 97%

Catalog No. AAL1394036
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1394036 500 g
AAL1394014 25 g
AAL1394022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1394036 Supplier Thermo Scientific Chemicals Supplier No. L1394036

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 124-43-6 | CH6N2O3 | 94.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Urea hydrogen peroxide is an antiseptic, disinfectant and bleaching agent used in pharmaceuticals and cosmetics respectively. It is used in the whitening of teeth and relieves minor inflammation of gums, oral mucosal surfaces and lips. It finds application in the preparation of plastics, blue print development and starch modification. In addition to this, it is used as a source of hydrogen peroxide easily handled in the laboratory.

Solubility
Soluble in water.

Notes
Incompatible with acetic anhydride, aniline and carboxylic acids.

Chemical Identifiers

CAS 124-43-6
Molecular Formula CH6N2O3
Molecular Weight (g/mol) 94.07
MDL Number MFCD00013119
InChI Key AQLJVWUFPCUVLO-UHFFFAOYSA-N
Synonym urea hydrogen peroxide, carbamide peroxide, percarbamide, urea peroxide, urea dioxide, hydrogen peroxide urea, urea hydroperoxide, hydroperit, hydroperite, percarbamid
PubChem CID 31294
ChEBI CHEBI:75178
IUPAC Name hydrogen peroxide;urea
SMILES C(=O)(N)N.OO

Specifications

Melting Point ∽90°C (decomposition)
Odor Odorless
Linear Formula H2NCONH2·H2O2
Quantity 500 g
UN Number UN1511
Beilstein 3680414
Sensitivity Very Hygroscopic
Merck Index 14,1782
Solubility Information Soluble in water.
Formula Weight 94.07
Percent Purity 97%
Chemical Name or Material Urea hydrogen peroxide adduct
Show More Show Less
Hazard Category H272-H314-H335
Hazard Statement GHS H Statement
H272-H314-H318
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: UREA HYDROGEN PEROXIDE
EINECSNumber 204-701-4
RTECSNumber T4860000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only