missing translation for 'onlineSavingsMsg'
Learn More

Urea, Crystal, ACS, 99.0-100.5%, Spectrum™ Chemical

Catalog No. 18614626
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614626 500 g Poly Bottle
18614625 2.5 kg Poly Pail
18614624 12 kg Poly Pail
18614627 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18614626 Supplier Spectrum Chemical Mfg Cor Supplier No. U1015500GM

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.06
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
IUPAC Name urea
SMILES NC(N)=O

Specifications

CAS Max % 1
Melting Point 132° to 135°C
Ignition Residue 0.0001
Packaging Poly Bottle
Quantity 500 g
Percent Purity 99.0 to 100.5%
Grade ACS
Insoluble Matter 0.0001

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.