missing translation for 'onlineSavingsMsg'
Learn More

Urea, BiotechGrade, 99-100.5%, Spectrum™ Chemical

Catalog No. 18607650
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607650 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18607650 Supplier Spectrum Chemical Mfg Cor Supplier No. U1024500GM
Only null left

U1024, 57-13-6, CH4N2O

Spectrum™ Chemical Urea, BiotechGrade is an odorless and colorless solid that is an important nitrogen-containing substance found in mammal urine. In concentrations up to 10 M, urea is a powerful protein denaturant as it disrupts the noncovalent bonds in the proteins. This property can be exploited to increase the solubility of some proteins. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.06
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
IUPAC Name urea
SMILES NC(N)=O

Specifications

CAS Max % 1
Melting Point 132° to 134°C
Ignition Residue 0.001
Odor Odorless
Packaging Poly Bottle
Quantity 500 g
Percent Purity 99.0 to 100.5%
Grade Biotech