missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Urapidil hydrochloride

Catalog No. p-7214761
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AC467590010 1 g
AC467590050 5 g
2 options

Catalog No. AC467590010

Supplier: Thermo Scientific Chemicals 467590010

Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64887-14-5
Molecular Formula C20H29N5O3·HCl

Specifications

Quantity 1 g
Melting Point 249.0°C to 250.0°C
Formula Weight 423.94
InChI Key KTMLZVUAXJERAT-UHFFFAOYSA-N
SMILES [H+].[Cl-].COC1=CC=CC=C1N1CCN(CCCNC2=CC(=O)N(C)C(=O)N2C)CC1
IUPAC Name hydrogen 6-({3-[4-(2-methoxyphenyl)piperazin-1-yl]propyl}amino)-1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione chloride
Molecular Weight (g/mol) 423.94
Chemical Name or Material Urapidil hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

RUO – Research Use Only