CAS RN 64887-14-5
IUPAC Name:
hydrogen 6-({3-[4-(2-methoxyphenyl)piperazin-1-yl]propyl}amino)-1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione chloride
Synonyms:
hydrogen urapidil chloride
Molecular Weight (g/mol):
423.94
Molecular Formula:
C20H30ClN5O3
InChi Key:
KTMLZVUAXJERAT-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].COC1=CC=CC=C1N1CCN(CCCNC2=CC(=O)N(C)C(=O)N2C)CC1
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Urapidil hydrochloride
Thermo Scientific Chemicals Urapidil hydrochloride
CAS: 64887-14-5 Molecular Formula: C20H30ClN5O3