missing translation for 'onlineSavingsMsg'
Learn More

Uranium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLU22X

 View more versions of this product

Catalog No. PLU2-2X


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
U1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Uranium Standard (Depleted)
7440-61-1 , 7697-37-2 , 7732-18-5
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
0.1,2.0,97.9
AA, ICP
Certified Reference Material
[U]
uranium
500 mL
CHEBI:49936
Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLU2-2Y, -2X    10,000    PLU2-3Y, -3X
Triple-leached LDPE bottle
Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
0.1,2.0,97.9
U
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
JFALSRSLKYAFGM-UHFFFAOYSA-N
2% HNO3
238.029
23989
Assurance

Chemical Identifiers

7440-61-1
238.029
23989
uranium
U
JFALSRSLKYAFGM-UHFFFAOYSA-N
CHEBI:49936
[U]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Harmful if swallowed. May be harmful if inhaled. Causes severe skin burns and eye damage

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only