missing translation for 'onlineSavingsMsg'
Learn More

Uranium ICP Standard, 1000 ppm U in 3% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company PU1KN100

Catalog No. PU1KN-100


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Uranium ICP Standard
1344-58-7,7697-37-2,7732-18-5
97.0,4.95,0.12
O3U
JCMLRUNDSXARRW-UHFFFAOYSA-N
trioxouranium
100 mL
1mL = 1mg U (1, 000ppm U)UO3 in 3% HNO3
Natural LDPE Bottle
93.0,4.76,0.11
Liquid
Yellow
O=[U](=O)=O
286.026
74013

Chemical Identifiers

1344-58-7
286.026
74013
O=[U](=O)=O
O3U
JCMLRUNDSXARRW-UHFFFAOYSA-N
trioxouranium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 18 months

SDS
Documents

Documents

Promotions

Promotions