missing translation for 'onlineSavingsMsg'
Learn More

Uracil, 99.75%, MP Biomedicals™

Catalog No. ICN10320425
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10320425 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10320425 Supplier MP Biomedicals Inc Supplier No. 0210320425

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66-22-8
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.09
MDL Number MFCD00006016
InChI Key ISAKRJDGNUQOIC-UHFFFAOYSA-N
Synonym uracil, 2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 2,4-dioxopyrimidine, pyrimidine-2,4 1h,3h-dione, pirod, 2,4-pyrimidinedione, pyrod, hybar x
PubChem CID 1174
ChEBI CHEBI:17568
IUPAC Name 1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES O=C1NC=CC(=O)N1

Specifications

Color White
Assay Percent Range 99.75%
Quantity 25 g
Formula Weight 112.09
Physical Form Crystalline Powder
Chemical Name or Material Uracil