missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Uracil, 99+%

Catalog No. AAA1557030
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1557030 250 g
AAA1557018 50 g
AAA155700B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1557030 Supplier Thermo Scientific Chemicals Supplier No. A1557030
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 66-22-8 | C4H4N2O2 | 112.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Uracil is produced by hydrolysis of nucleic acid and used in biochemical research. It serves as a allosteric regulator and coenzyme for reactions in the human body and in plants. It reacts with fluorine to get 5-fluorouracil, which is an anticancer drug. Further, it is used in drug delivery and in pharmaceuticals. In addition to this, it is used in the synthesis of caffeine and in the determination of microbial contamination of tomatoes.

Solubility
Soluble in water, aqueous ammonia, 1M sodium hydroxide solution and alkalies. Insoluble in ethanol and ether.

Notes
Incompatible with water, air and strong oxidizing agents.

Chemical Identifiers

CAS 66-22-8
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.09
MDL Number MFCD00006016
InChI Key ISAKRJDGNUQOIC-UHFFFAOYSA-N
Synonym uracil, 2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 2,4-dioxopyrimidine, pyrimidine-2,4 1h,3h-dione, pirod, 2,4-pyrimidinedione, pyrod, hybar x
PubChem CID 1174
ChEBI CHEBI:17568
IUPAC Name 1H-pyrimidine-2,4-dione
SMILES O=C1NC=CC(=O)N1

Specifications

Melting Point ∼330°C
Assay Percent Range ≥99%
Quantity 250 g
Beilstein 606623
Merck Index 14,9850
Solubility Information Soluble in water,aqueous ammonia,1M sodium hydroxide solution and alkalies. Insoluble in ethanol and ether.
Formula Weight 112.09
Percent Purity ≥99%
Chemical Name or Material Uracil
EINECSNumber 200-621-9
RTECSNumber YQ8650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only