missing translation for 'onlineSavingsMsg'
Learn More

Uracil-5-boronic acid, 95%

Catalog No. AAH2721903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2721903 1 g
AAH27219MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2721903 Supplier Thermo Scientific Chemicals Supplier No. H2721903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 70523-22-7 | C4H5BN2O4 | 155.90 g/mol

It finds its application in the probing the interactions between boronic acids and cis-diol-containing biomolecules by affinity capillary electrophoresis. It is also used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the probing the interactions between boronic acids and cis-diol-containing biomolecules by affinity capillary electrophoresis. It is also used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 70523-22-7
Molecular Formula C4H5BN2O4
Molecular Weight (g/mol) 155.90
MDL Number MFCD01318983
InChI Key PVEJOCQTIVCDNO-UHFFFAOYSA-N
Synonym uracil-5-boronic acid, 2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl boronic acid, 2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinylboronic acid, 2,4-dihydroxypyrimidine-5-boronic acid, 2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-ylboronic acid, 2,4-dihydroxypyrimidin-5-ylboronic acid, 2,4-dioxo-1h-pyrimidin-5-yl boronic acid, 2,4-dioxo-1,3-dihydropyrimidin-5-ylboronic acid, t6mvmvj ebqq wln, 5-dihydroxyboryluracil
PubChem CID 256205
IUPAC Name (2,4-dioxo-1H-pyrimidin-5-yl)boronic acid
SMILES OB(O)C1=CNC(=O)NC1=O

Specifications

Melting Point >300°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 155.9
Percent Purity 95%
Chemical Name or Material Uracil-5-boronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only