Learn More
Idex Threaded Plugs and Caps
Plugs to close off unused ports in valves and multi-port connectors
- Seal 6-32, 10-32, 1/4-28, M6x1 and 5/16-24 threaded port
$6.83 - $10.28
| Catalog Number | Mfr. No. | Type | Color | Material | Port Type | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | Color | Material | Port Type | Price | Quantity | ||
05-700-111
|
Idex
UPP311 |
Plug | Natural | Tefzel | 1/4-28 Flat Bottom |
Each for $7.57
|
|
||
05-700-114
|
Idex
UPP314 |
Plug | Black | Tefzel | M6 Flat Bottom |
Each for $6.83
|
|
||
05-700-190
|
Idex
UPP754 |
Cap | Yellow | Tefzel | 5/16-24 |
Each for $10.19
|
|
||
05-700-192
|
Idex
UPP756 |
Cap | Blue | Tefzel | M6 Flat Bottom |
Each for $10.28
|
|
||
05-700-191
|
Idex
UPP755DUP |
Cap | Black | Tefzel | 1/4-28 Flat Bottom |
Each for $9.85
|
|
||
05-703-13
|
Idex
UPP550 |
Plug | Natural | PEEK | 10-32 Coned |
Each for $9.17
|
|
||
29-008-623
|
Idex
UPP316 |
Plug | Natural | PFA | 1/4-28 Flat Bottom |
Each for $7.38
|
|
||
Description
Plugs to close off unused ports in valves and multi-port connectors
- Seal 6-32, 10-32, 1/4-28, M6x1 and 5/16-24 threaded port