Learn More
Idex Threaded Plugs and Caps
Plugs to close off unused ports in valves and multi-port connectors
- Seal 6-32, 10-32, 1/4-28, M6x1 and 5/16-24 threaded port
Supplier: Idex UPP550
Description
Plugs to close off unused ports in valves and multi-port connectors
- Seal 6-32, 10-32, 1/4-28, M6x1 and 5/16-24 threaded port
Specifications
| Plug | |
| Natural | |
| 10-32 Coned | |
| PEEK |