Learn More
Idex Threaded Plugs and Caps DFS Item
Plugs to close off unused ports in valves and multi-port connectors
  • Seal 6-32, 10-32, 1/4-28, M6x1 and 5/16-24 threaded port
Supplier:  Idex UPP755DUP
 View more versions of this product
Catalog No. 05-700-191
Only null left
Explore available promotions
Description
Description
Plugs to close off unused ports in valves and multi-port connectors
  • Seal 6-32, 10-32, 1/4-28, M6x1 and 5/16-24 threaded port
Specifications
Specifications
Cap
Black
1/4-28 Flat Bottom
Tefzel
Product Suggestions
Product Suggestions